siRNA Details
Gene Symbol: HIPK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144685.4 | NP_653286.2 | 1 | 711 | ||
XM_006723036.2 | XP_006723099.1 | 1 | 686 | |||
GenBank mRNA | AK057380.1 | - | 1 | 686 | ||
AK292228.1 | - | 1 | 672 | |||
AK301713.1 | - | 2 | 464 | |||
BC034501.2 | - | 1 | 677 | 4823304 |
