siRNA Details
Gene Symbol: MYCL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005376.4 | NP_005367.2 | 2 | 1800 | ||
GenBank mRNA | BC011864.2 | - | 2 | 1688 | 4541675 | |
BM967204.1 | - | 1 | 209 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
