siRNA Details
Gene Symbol: EPM2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005670.3 | NP_005661.1 | 4 | 2847 | ||
XM_006715564.3 | XP_006715627.1 | 3 | 2768 | |||
XM_011536116.1 | XP_011534418.1 | 4 | 2223 | |||
XM_017011300.1 | XP_016866789.1 | 4 | 2350 | |||
XM_017011301.1 | XP_016866790.1 | 4 | 2440 | |||
XM_017011302.1 | XP_016866791.1 | 4 | 2195 | |||
GenBank mRNA | AF084535.2 | - | 4 | 2497 | ||
AF284580.1 | - | 4 | 2849 | |||
AF454491.1 | - | 4 | 2266 | |||
AF454492.1 | - | 4 | 3033 | |||
AF454493.1 | - | 5 | 2737 | |||
AJ130763.1 | - | 4 | 2253 | |||
AK022721.1 | - | 3 | 2112 | |||
BC070047.1 | - | 4 | 2514 | 5287405 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
