siRNA Details
Gene Symbol: MBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025081.1 | NP_001020252.1 | 6 | 597 | ||
NM_001025090.1 | NP_001020261.1 | 5 | 519 | |||
NM_001025092.1 | NP_001020263.1 | 4 | 486 | |||
NM_001025101.1 | NP_001020272.1 | 8 | 1091 | |||
NM_002385.2 | NP_002376.1 | 5 | 564 | |||
GenBank mRNA | AK098402.1 | - | 5 | 475 | ||
AK122594.1 | - | 1 | 1042 | |||
AK123433.1 | - | 1 | 3139 | |||
AK124830.1 | - | 1 | 1816 | |||
AK128770.1 | - | 5 | 478 | |||
AK128788.1 | - | 5 | 475 | |||
AK289893.1 | - | 5 | 519 | |||
AK293922.1 | - | 4 | 514 | |||
AK314553.1 | - | 6 | 593 | |||
BC008749.2 | - | 5 | 457 | 3346521 | ||
BC068550.1 | - | 3 | 333 | |||
BC080654.1 | - | 4 | 431 | 4859683 | ||
BC101771.1 | - | 5 | 457 | 8069277 | ||
BC101773.1 | - | 5 | 457 | 8069279 | ||
BC143348.1 | - | 4 | 424 | 9051853 | ||
BC143350.1 | - | 5 | 457 | 9051855 | ||
BG766942.1 | - | 5 | 519 | |||
CB153535.1 | - | 6 | 484 | |||
CB156561.1 | - | 5 | 392 | |||
CR536534.1 | - | 4 | 395 | |||
CR541919.1 | - | 4 | 395 | |||
CR627018.1 | - | 4 | 3946 | |||
M13577.1 | - | 5 | 464 | |||
M30047.1 | - | 4 | 427 | |||
M30515.1 | - | 6 | 513 | |||
M30516.1 | - | 5 | 483 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
