siRNA Details
Gene Symbol: HOXC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017410.2 | NP_059106.2 | 2 | 2034 | ||
GenBank mRNA | AF263466.2 | - | 2 | 1990 | ||
AK024027.1 | - | 2 | 1516 | |||
BC002754.1 | - | 3 | 1935 | |||
BC090850.1 | - | 2 | 2003 | 6171228 |
