siRNA Details
Gene Symbol: STYX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130701.1 | NP_001124173.1 | 11 | 584 | ||
NM_145251.3 | NP_660294.1 | 10 | 832 | |||
XM_006720242.3 | XP_006720305.1 | 8 | 452 | |||
XM_011537108.2 | XP_011535410.1 | 10 | 499 | |||
XM_011537109.2 | XP_011535411.1 | 7 | 393 | |||
GenBank mRNA | AK098195.1 | - | 10 | 815 | ||
AL832591.1 | - | 11 | 569 | |||
BC020265.2 | - | 10 | 783 | 3922829 | ||
BC146995.1 | - | 10 | 722 | 9056923 | ||
BC146998.1 | - | 10 | 722 | 9056926 | ||
BX647636.1 | - | 10 | 790 | |||
U87169.1 | - | 4 | 178 |
