siRNA Details
Gene Symbol: PTPDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253829.1 | NP_001240758.1 | 3, 4 | 607 | ||
NM_001253830.1 | NP_001240759.1 | 4, 5 | 668 | |||
NM_152422.4 | NP_689635.3 | 3, 4 | 601 | |||
NM_177995.2 | NP_818931.1 | 4, 5 | 683 | |||
XM_005251711.4 | XP_005251768.1 | 4, 5 | 2625 | |||
XM_011518240.2 | XP_011516542.1 | 4, 5 | 433 | |||
XM_011518241.2 | XP_011516543.1 | 4, 5 | 387 | |||
XM_017014280.1 | XP_016869769.1 | 5, 6 | 492 | |||
XM_017014281.1 | XP_016869770.1 | 4, 5 | 411 | |||
GenBank mRNA | AK124912.1 | - | 3, 4 | 487 | ||
AK289692.1 | - | 3, 4 | 558 | |||
AK310403.1 | - | 3, 4 | 565 | |||
AY171233.1 | - | 4, 5 | 688 | |||
BC067120.1 | - | 4, 5 | 668 | 5539182 | ||
DA312531.1 | - | 2, 3 | 183 |
