siRNA Details
Gene Symbol: PLK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005030.5 | NP_005021.2 | 1 | 532 | ||
GenBank mRNA | AB084459.1 | - | 1 | 375 | ||
AB209179.1 | - | 1 | 1648 | |||
AK303263.1 | - | 2 | 148 | |||
AK308276.1 | - | 2 | 390 | |||
AK313227.1 | - | 1 | 456 | |||
BC002369.2 | - | 1 | 424 | 2822226 | ||
BC003002.1 | - | 1 | 402 | 2823623 | ||
BC014846.1 | - | 1 | 377 | 3854860 | ||
L19559.1 | - | 1 | 434 | |||
U01038.1 | - | 1 | 434 | |||
X73458.1 | - | 1 | 448 | |||
X75932.1 | - | 1 | 434 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
