siRNA Details
Gene Symbol: DLX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178120.4 | NP_835221.2 | 2 | 638 | ||
GenBank mRNA | AK095266.1 | - | 1 | 1268 | ||
AK290503.1 | - | 2 | 611 | |||
AK308842.1 | - | 2 | 561 | |||
AY257976.1 | - | 2 | 638 | |||
BC036189.1 | - | 2 | 604 | 5311747 | ||
KJ534824.1 | - | 2 | 440 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
