siRNA Details
Gene Symbol: LSMEM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134468.1 | NP_001127940.1 | 4 | 823 | ||
NM_182597.2 | NP_872403.1 | 4 | 1029 | |||
XM_011516074.1 | XP_011514376.1 | 4 | 1532 | |||
XM_011516075.2 | XP_011514377.1 | 4 | 3219 | |||
XM_011516076.2 | XP_011514378.1 | 4 | 2855 | |||
XM_017012028.1 | XP_016867517.1 | 3 | 2737 | |||
GenBank mRNA | AK096894.1 | - | 4 | 1045 | ||
BC126149.1 | - | 4 | 867 | 8991865 | ||
BC130568.1 | - | 4 | 867 | 40146599 |
