siRNA Details
Gene Symbol: BACH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001186.3 | NP_001177.1 | 5 | 2461 | ||
NM_206866.2 | NP_996749.1 | 5 | 2485 | |||
GenBank mRNA | AB002803.1 | - | 5 | 2360 | ||
AK291021.1 | - | 5 | 2418 | |||
BC063307.1 | - | 5 | 2334 | 30340811 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
