siRNA Details
Gene Symbol: SMARCB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003073.4 | NP_003064.2 | 2, 3 | 467 | ||
XM_011530345.2 | XP_011528647.1 | 2, 3 | 436 | |||
GenBank mRNA | AJ011737.1 | - | 2, 3 | 375 | ||
AK299411.1 | - | 2, 3 | 397 | |||
AL389878.1 | - | 2, 3 | 277 | |||
AY207471.1 | - | 1, 2 | 80 | |||
CR456581.1 | - | 2, 3 | 281 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
