siRNA Details
Gene Symbol: TRIM21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003141.3 | NP_003132.2 | 2 | 346 | ||
GenBank mRNA | AK225564.1 | - | 2 | 329 | ||
AY742713.1 | - | 1 | 233 | |||
BC010861.1 | - | 2 | 317 | 3900540 | ||
BP280435.1 | - | 2 | 346 | |||
BP288941.1 | - | 2 | 327 | |||
M34551.1 | - | 1 | 271 | |||
M62800.1 | - | 2 | 318 |
