siRNA Details
Gene Symbol: ZNF135 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164529.1 | NP_001158001.1 | 4 | 370 | ||
NM_001164530.1 | NP_001158002.1 | 4 | 729 | |||
NM_001289401.1 | NP_001276330.1 | 5 | 796 | |||
NM_001289402.1 | NP_001276331.1 | 5 | 1159 | |||
NM_003436.3 | NP_003427.3 | 5 | 827 | |||
NM_007134.1 | NP_009065.1 | 4 | 765 | |||
XM_005259211.1 | XP_005259268.1 | 4 | 729 | |||
XM_006723362.3 | XP_006723425.1 | 5 | 835 | |||
XM_006723363.3 | XP_006723426.1 | 5 | 1199 | |||
XM_017027240.1 | XP_016882729.1 | 4 | 1152 | |||
XM_017027241.1 | XP_016882730.1 | 5 | 1154 | |||
XM_017027242.1 | XP_016882731.1 | 4 | 594 | |||
GenBank mRNA | AK094202.1 | - | 5 | 1118 | ||
AK098011.1 | - | 4 | 729 | |||
AK295110.1 | - | 5 | 797 | |||
AK297226.1 | - | 4 | 560 | |||
AL833637.1 | - | 4 | 328 | |||
BC046434.1 | - | 5 | 791 | 4824328 | ||
BX647775.1 | - | 4 | 733 | |||
U09413.1 | - | 1 | 198 |
