siRNA Details
Gene Symbol: TP53BP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031685.2 | NP_001026855.2 | 13 | 2521 | ||
NM_005426.2 | NP_005417.1 | 14 | 2653 | |||
XM_011544268.1 | XP_011542570.1 | 12 | 2382 | |||
XM_011544269.2 | XP_011542571.1 | 10 | 2040 | |||
XM_017002220.1 | XP_016857709.1 | 14 | 2662 | |||
GenBank mRNA | AJ318888.2 | - | 13 | 2466 | ||
AK294432.1 | - | 13 | 2373 | |||
AK294654.1 | - | 3 | 457 | |||
AK294964.1 | - | 6 | 1541 | |||
AK295378.1 | - | 7 | 1245 | |||
AK316016.1 | - | 6 | 1443 | |||
AK316103.1 | - | 6 | 1421 | |||
BC040247.1 | - | 11 | 1997 | 4454266 | ||
BC058918.1 | - | 13 | 2479 | 4537206 | ||
U09582.1 | - | 2 | 416 | |||
U58334.1 | - | 14 | 2598 |
