siRNA Details
Gene Symbol: PURG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323311.1 | NP_001310240.1 | 2 | 1014 | ||
NM_013357.2 | NP_037489.1 | 1 | 910 | |||
GenBank mRNA | AF195513.2 | - | 1 | 1824 | ||
BC106708.2 | - | 1 | 910 | 40005076 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
