siRNA Details
Gene Symbol: ZNF354C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014594.2 | NP_055409.1 | 5 | 591 | ||
XM_017009409.1 | XP_016864898.1 | 5 | 694 | |||
GenBank mRNA | AF395540.1 | - | 3 | 258 | ||
AF525463.1 | - | 5 | 400 | |||
BC063312.1 | - | 5 | 591 | 30341631 | ||
CN482682.1 | - | 5 | 569 |
