siRNA Details
Gene Symbol: ZNF777 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015694.2 | NP_056509.2 | 4 | 1162 | ||
XM_005249980.3 | XP_005250037.1 | 4 | 1148 | |||
XM_011516055.2 | XP_011514357.1 | 4 | 1172 | |||
GenBank mRNA | AB033111.1 | - | 4 | 1076 | ||
AK074515.1 | - | 2 | 48 | |||
AK075032.1 | - | 3 | 259 | |||
BC023985.2 | - | 3 | 226 | 2820826 | ||
BC105968.1 | - | 3 | 496 |
