siRNA Details
Gene Symbol: ZNF586 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077426.2 | NP_001070894.1 | 2 | 276 | ||
NM_001204814.1 | NP_001191743.1 | 4 | 453 | |||
NM_017652.3 | NP_060122.2 | 3 | 403 | |||
GenBank mRNA | AB074270.1 | - | 3 | 386 | ||
AK000077.1 | - | 3 | 385 | |||
AK095993.1 | - | 4 | 451 | |||
AK300419.1 | - | 2 | 233 | |||
AK302151.1 | - | 4 | 359 | |||
BC124558.1 | - | 2 | 214 | 40085152 | ||
BP352135.1 | - | 2 | 277 | |||
DA904413.1 | - | 3 | 398 |
