siRNA Details
Gene Symbol: ZNF416 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017879.1 | NP_060349.1 | 4 | 478 | ||
XM_005259051.3 | XP_005259108.1 | 3 | 414 | |||
GenBank mRNA | AK000564.1 | - | 4 | 530 | ||
BC000130.1 | - | 4 | 478 | 2901256 |
