siRNA Details
Gene Symbol: UCKL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193379.1 | NP_001180308.1 | 3, 4 | 722 | ||
NM_017859.3 | NP_060329.2 | 3, 4 | 494 | |||
NR_126526.1 | 3, 4 | 494 | ||||
XM_006723809.1 | XP_006723872.1 | 3, 4 | 494 | |||
XM_006723810.1 | XP_006723873.1 | 3, 4 | 491 | |||
XM_017027896.1 | XP_016883385.1 | 3, 4 | 494 | |||
XM_017027897.1 | XP_016883386.1 | 3, 4 | 494 | |||
XM_017027898.1 | XP_016883387.1 | 3, 4 | 491 | |||
XM_017027899.1 | XP_016883388.1 | 3, 4 | 491 | |||
XM_017027900.1 | XP_016883389.1 | 3, 4 | 1340 | |||
XM_017027901.1 | XP_016883390.1 | 3, 4 | 1338 | |||
XM_017027902.1 | XP_016883391.1 | 4, 5 | 694 | |||
GenBank mRNA | AJ605558.1 | - | 3, 4 | 463 | ||
AK000524.1 | - | 3, 4 | 463 | |||
AK222559.1 | - | 3, 4 | 465 | |||
AK223220.1 | - | 3, 4 | 463 | |||
AK303688.1 | - | 3, 4 | 674 | |||
BC033078.1 | - | 3, 4 | 432 | 4810505 | ||
DA255371.1 | - | 3, 4 | 494 | |||
DB193745.1 | - | 3, 4 | 494 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
