siRNA Details
Gene Symbol: ZNF286A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130842.1 | NP_001124314.1 | 5 | 696 | ||
NM_001288642.1 | NP_001275571.1 | 5 | 623 | |||
NM_001288643.1 | NP_001275572.1 | 4 | 464 | |||
NM_001288644.1 | NP_001275573.1 | 3 | 349 | |||
NM_001288645.1 | NP_001275574.1 | 5 | 783 | |||
NM_001288646.1 | NP_001275575.1 | 5 | 696 | |||
NM_001288647.1 | NP_001275576.1 | 4 | 581 | |||
NM_001288648.1 | NP_001275577.1 | 4 | 464 | |||
NM_001288649.1 | NP_001275578.1 | 3 | 285 | |||
NM_020652.2 | NP_065703.1 | 5 | 783 | |||
GenBank mRNA | AF086305.1 | - | 3 | 216 | ||
AF217226.1 | - | 4 | 615 | |||
AK092259.1 | - | 5 | 621 | |||
AK315062.1 | - | 5 | 541 | |||
BC015730.1 | - | 5 | 621 | |||
BC025260.1 | - | 5 | 621 | |||
BC128259.1 | - | 4 | 451 | 40116891 | ||
BC128411.1 | - | 4 | 451 | 40116881 | ||
DA096356.1 | - | 4 | 287 | |||
DA188283.1 | - | 4 | 439 | |||
DA748526.1 | - | 5 | 534 | |||
DA786711.1 | - | 3 | 123 | |||
DA796080.1 | - | 5 | 456 | |||
DA806764.1 | - | 3 | 197 | |||
DC344287.1 | - | 4 | 350 | |||
DN997307.1 | - | 4 | 479 |
