siRNA Details
Gene Symbol: TRIM34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003827.1 | NP_001003827.1 | 2 | 425 | ||
NM_021616.5 | NP_067629.2 | 2 | 429 | |||
NM_130390.2 | NP_569074.2 | 1 | 345 | |||
GenBank mRNA | AB039902.1 | - | 2 | 433 | ||
AB039904.1 | - | 1 | 327 | |||
AF220143.1 | - | 2 | 412 | |||
AF220144.1 | - | 2 | 412 | |||
AK027876.1 | - | 2 | 401 | |||
AL583914.1 | - | 1 | 34 | |||
DB024660.1 | - | 2 | 429 |
Gene Symbol: TRIM6-TRIM34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003819.3 | NP_001003819.1 | 8 | 1593 | ||
GenBank mRNA | AB039903.1 | - | 8 | 1501 | ||
BC136871.1 | - | 8 | 1512 | 9020862 | ||
BC140722.1 | - | 8 | 1512 | 9021590 | ||
BC144424.1 | - | 8 | 1512 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
