siRNA Details
Gene Symbol: TRIM34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003827.1 | NP_001003827.1 | 4 | 732 | ||
NM_021616.5 | NP_067629.2 | 4 | 736 | |||
NM_130390.2 | NP_569074.2 | 3 | 652 | |||
GenBank mRNA | AB039902.1 | - | 4 | 740 | ||
AB039904.1 | - | 3 | 634 | |||
AF220143.1 | - | 4 | 719 | |||
AF220144.1 | - | 5 | 652 | |||
AK027876.1 | - | 4 | 708 | |||
AL583914.1 | - | 3 | 341 |
Gene Symbol: TRIM6-TRIM34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003819.3 | NP_001003819.1 | 10 | 1900 | ||
GenBank mRNA | AB039903.1 | - | 10 | 1808 | ||
BC136871.1 | - | 10 | 1819 | 9020862 | ||
BC140722.1 | - | 10 | 1819 | 9021590 | ||
BC144424.1 | - | 10 | 1819 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
