siRNA Details
Gene Symbol: ZNF350 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021632.3 | NP_067645.3 | 4, 5 | 465 | ||
XM_011527196.1 | XP_011525498.1 | 4, 5 | 509 | |||
XM_017027098.1 | XP_016882587.1 | 5, 6 | 742 | |||
XM_017027099.1 | XP_016882588.1 | 6, 7 | 765 | |||
XM_017027100.1 | XP_016882589.1 | 4, 5 | 624 | |||
GenBank mRNA | AF295096.1 | - | 3, 4 | 420 | ||
AF309561.1 | - | 4, 5 | 454 | |||
AK023467.1 | - | 4, 5 | 465 | |||
AU135269.1 | - | 4, 5 | 465 | |||
BC009921.2 | - | 3, 4 | 405 | 3506774 |
