siRNA Details
Gene Symbol: RFWD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286644.1 | NP_001273573.1 | 5 | 950 | ||
NM_022457.6 | NP_071902.2 | 7 | 1147 | |||
XM_005245447.2 | XP_005245504.1 | 7 | 1170 | |||
XM_006711487.2 | XP_006711550.1 | 6 | 1113 | |||
XM_011509881.1 | XP_011508183.1 | 7 | 1182 | |||
XM_017002059.1 | XP_016857548.1 | 7 | 1182 | |||
XM_017002060.1 | XP_016857549.1 | 7 | 1170 | |||
XM_017002061.1 | XP_016857550.1 | 7 | 1182 | |||
XM_017002063.1 | XP_016857552.1 | 7 | 1170 | |||
XM_017002064.1 | XP_016857553.1 | 6 | 1113 | |||
XM_017002066.1 | XP_016857555.1 | 6 | 1113 | |||
XM_017002070.1 | XP_016857559.1 | 6 | 888 | |||
XM_017002071.1 | XP_016857560.1 | 4 | 277 | |||
XM_017002072.1 | XP_016857561.1 | 4 | 275 | |||
XM_017002076.1 | XP_016857565.1 | 5 | 334 | |||
XM_017002077.1 | XP_016857566.1 | 2 | 72 | |||
GenBank mRNA | AF508940.1 | - | 7 | 881 | ||
AF527539.1 | - | 7 | 1144 | |||
AK314750.1 | - | 7 | 1079 | |||
BC039723.1 | - | 5 | 397 | |||
BC094728.1 | - | 7 | 1135 | 5271629 | ||
KJ534928.1 | - | 7 | 881 | |||
KJ534931.1 | - | 6 | 812 | |||
KJ534932.1 | - | 7 | 869 | |||
KJ535061.1 | - | 4 | 586 | |||
KJ535076.1 | - | 6 | 783 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
