siRNA Details
Gene Symbol: EBF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022659.3 | NP_073150.2 | 10, 11 | 1282 | ||
GenBank mRNA | AK021562.1 | - | 3, 4 | 321 | ||
AK304372.1 | - | 9, 10 | 930 | |||
AY700779.1 | - | 2, 3 | 210 | |||
BC069665.1 | - | 3, 4 | 299 | |||
BC069726.1 | - | 3, 4 | 302 | |||
BC069747.1 | - | 3, 4 | 301 | |||
BC069768.1 | - | 3, 4 | 302 | |||
BC074794.2 | - | 2, 3 | 238 | |||
BC113478.1 | - | 2, 3 | 266 | |||
BC113504.1 | - | 2, 3 | 266 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
