siRNA Details
Gene Symbol: ZNF606 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025027.3 | NP_079303.2 | 7 | 1232 | ||
XM_005259276.1 | XP_005259333.1 | 7 | 1092 | |||
GenBank mRNA | AB058755.1 | - | 7 | 1235 | ||
AF455357.1 | - | 8 | 1794 | |||
AK293097.1 | - | 7 | 702 | |||
AK294037.1 | - | 6 | 616 | |||
BC037209.1 | - | 7 | 706 | 4837545 |
