siRNA Details
Gene Symbol: ZNF397 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135178.2 | NP_001128650.1 | 3 | 640 | ||
NM_032347.2 | NP_115723.1 | 3 | 640 | |||
XM_006722558.2 | XP_006722621.1 | 3 | 693 | |||
XM_011526229.2 | XP_011524531.1 | 4 | 782 | |||
XM_011526230.2 | XP_011524532.1 | 3 | 984 | |||
XM_011526231.2 | XP_011524533.1 | 3 | 581 | |||
XM_017026042.1 | XP_016881531.1 | 3 | 636 | |||
GenBank mRNA | AF533250.2 | - | 3 | 633 | ||
AK027384.1 | - | 3 | 615 | |||
AK311533.1 | - | 2 | 125 | |||
AY157641.2 | - | 3 | 633 | |||
BC006172.1 | - | 3 | 615 | 4053312 |
