siRNA Details
Gene Symbol: ZNF397 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135178.2 | NP_001128650.1 | 4 | 796 | ||
NM_032347.2 | NP_115723.1 | 4 | 796 | |||
XM_006722558.2 | XP_006722621.1 | 4 | 849 | |||
XM_011526229.2 | XP_011524531.1 | 5 | 938 | |||
XM_011526230.2 | XP_011524532.1 | 4 | 1140 | |||
XM_011526231.2 | XP_011524533.1 | 4 | 737 | |||
XM_017026042.1 | XP_016881531.1 | 4 | 792 | |||
GenBank mRNA | AF533250.2 | - | 4 | 789 | ||
AK311533.1 | - | 3 | 281 | |||
BC006172.1 | - | 4 | 771 | 4053312 |
