siRNA Details
Gene Symbol: ZNF101 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300949.1 | NP_001287878.1 | 4 | 753 | ||
NM_033204.3 | NP_149981.2 | 4 | 881 | |||
GenBank mRNA | AK097169.1 | - | 4 | 873 | ||
AL096747.1 | - | 3 | 2426 | |||
AY149174.1 | - | 4 | 815 | |||
BC011502.1 | - | 1 | 517 | |||
BC094808.1 | - | 4 | 814 | |||
BC119680.1 | - | 5 | 917 | 40116294 |
