siRNA Details
Gene Symbol: REG3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002580.2 | NP_002571.1 | 2, 3 | 138 | ||
NM_138937.2 | NP_620354.1 | 2, 3 | 115 | |||
NM_138938.2 | NP_620355.1 | 1, 2 | 425 | |||
GenBank mRNA | AK291973.1 | - | 2, 3 | 114 | ||
AY544128.1 | - | 1, 2 | 162 | |||
AY544130.1 | - | 1, 2 | 108 | |||
BC036776.1 | - | 1, 2 | 119 | 5749980 | ||
BQ777112.1 | - | 2, 3 | 138 | |||
D13510.1 | - | 1, 2 | 114 | |||
DC300833.1 | - | 1, 2 | 426 | |||
M84337.1 | - | 1, 2 | 119 | |||
S51768.1 | - | 1, 2 | 119 | |||
X68641.1 | - | 1, 2 | 90 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
