siRNA Details
Gene Symbol: ZNF584 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318002.1 | NP_001304931.1 | 2 | 526 | ||
NM_173548.2 | NP_775819.1 | 2 | 550 | |||
XM_005258628.4 | XP_005258685.1 | 2 | 1029 | |||
XM_005258630.4 | XP_005258687.1 | 2 | 544 | |||
GenBank mRNA | AK290068.1 | - | 2 | 535 | ||
AK298929.1 | - | 2 | 423 | |||
BC033149.1 | - | 2 | 528 | 4125244 | ||
DC350090.1 | - | 2 | 517 |
