siRNA Details
Gene Symbol: PDE9A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001567.1 | NP_001001567.1 | 1, 2 | 140 | ||
NM_001001570.1 | NP_001001570.1 | 1, 2 | 140 | |||
NM_001001574.1 | NP_001001574.1 | 1, 2 | 140 | |||
NM_001001579.1 | NP_001001579.1 | 1, 2 | 140 | |||
NM_001001583.1 | NP_001001583.1 | 1, 2 | 140 | |||
NM_001315533.1 | NP_001302462.1 | 1, 2 | 140 | |||
NM_002606.2 | NP_002597.1 | 1, 2 | 140 | |||
XM_011529600.2 | XP_011527902.1 | 1, 2 | 113 | |||
XM_017028366.1 | XP_016883855.1 | 1, 2 | 113 | |||
XM_017028367.1 | XP_016883856.1 | 1, 2 | 113 | |||
GenBank mRNA | AF048837.1 | - | 1, 2 | 137 | ||
AF067223.1 | - | 1, 2 | 89 | |||
AF067224.1 | - | 1, 2 | 88 | |||
AK001307.1 | - | 1, 2 | 111 | |||
AK314679.1 | - | 1, 2 | 113 | |||
AY196299.1 | - | 1, 2 | 113 | |||
AY196303.1 | - | 1, 2 | 113 | |||
AY196308.1 | - | 1, 2 | 113 | |||
AY196312.1 | - | 1, 2 | 113 | |||
AY701187.1 | - | 1, 2 | 137 | |||
BC009047.1 | - | 1, 2 | 114 | 3874635 | ||
BT007016.1 | - | 1, 2 | 77 | |||
KU178246.1 | - | 1, 2 | 77 | |||
KU178247.1 | - | 1, 2 | 77 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
