siRNA Details
Gene Symbol: ALAS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000032.4 | NP_000023.2 | 4 | 484 | ||
XM_005261995.3 | XP_005262052.1 | 5 | 504 | |||
XM_017029354.1 | XP_016884843.1 | 5 | 505 | |||
GenBank mRNA | AK290565.1 | - | 3 | 357 | ||
AK313118.1 | - | 4 | 484 | |||
BP233279.1 | - | 5 | 504 | |||
X56352.1 | - | 3 | 343 | |||
X60364.1 | - | 4 | 398 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
