siRNA Details
Gene Symbol: SLC11A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000578.3 | NP_000569.3 | 15 | 2713 | ||
XM_005246793.3 | XP_005246850.1 | 14 | 2833 | |||
XM_005246794.3 | XP_005246851.1 | 16 | 3050 | |||
XM_006712709.3 | XP_006712772.1 | 15 | 2907 | |||
XM_006712710.3 | XP_006712773.1 | 14 | 2784 | |||
XM_006712711.3 | XP_006712774.1 | 13 | 2710 | |||
XM_011511684.2 | XP_011509986.1 | 16 | 3085 | |||
XM_011511685.2 | XP_011509987.1 | 15 | 2942 | |||
XM_017004765.1 | XP_016860254.1 | 14 | 2853 | |||
XM_017004766.1 | XP_016860255.1 | 15 | 2944 | |||
XM_017004767.1 | XP_016860256.1 | 12 | 2171 | |||
GenBank mRNA | AB209650.1 | - | 13 | 5293 | ||
BC033754.1 | - | 14 | 2147 | |||
D38171.1 | - | 13 | 2225 | |||
D50402.1 | - | 15 | 2553 | |||
D50403.1 | - | 15 | 2464 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
