siRNA Details
Gene Symbol: CSNK1G2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319.6 | NP_001310.3 | 12 | 2048 | ||
XM_005259498.1 | XP_005259555.1 | 11 | 2150 | |||
XM_005259499.3 | XP_005259556.1 | 11 | 1957 | |||
XM_005259500.1 | XP_005259557.1 | 11 | 2135 | |||
XM_005259501.2 | XP_005259558.1 | 11 | 2509 | |||
XM_017026297.1 | XP_016881786.1 | 11 | 2024 | |||
XM_017026298.1 | XP_016881787.1 | 12 | 2434 | |||
GenBank mRNA | AY007134.1 | - | 6 | 749 | ||
BC020972.1 | - | 11 | 1578 | 3851613 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
