siRNA Details
Gene Symbol: MYL6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199629.1 | NP_001186558.1 | 6, 7 | 927 | ||
NM_002475.4 | NP_002466.1 | 6, 7 | 863 | |||
GenBank mRNA | BC012425.1 | - | 5, 6 | 651 | 3862144 | |
BC014400.1 | - | 6, 7 | 927 | 3503030 | ||
M31211.1 | - | 5, 6 | 631 | |||
X16434.1 | - | 5, 6 | 616 |
