siRNA Details
Gene Symbol: SCP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007098.2 | NP_001007099.1 | 3 | 350 | ||
NM_001193600.1 | NP_001180529.1 | 3 | 350 | |||
NM_001193617.1 | NP_001180546.1 | 2 | 292 | |||
NM_002979.4 | NP_002970.2 | 3 | 350 | |||
XM_005271103.4 | XP_005271160.1 | 3 | 293 | |||
XM_011541935.2 | XP_011540237.1 | 3 | 293 | |||
XM_017002045.1 | XP_016857534.1 | 3 | 293 | |||
GenBank mRNA | AB208789.1 | - | 3 | 302 | ||
AK295214.1 | - | 2 | 235 | |||
AK308105.1 | - | 3 | 293 | |||
AK312856.1 | - | 3 | 301 | |||
BC067108.1 | - | 3 | 261 | 5176900 | ||
BG432929.1 | - | 304 | ||||
DC330171.1 | - | 3 | 334 | |||
M75883.1 | - | 3 | 203 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
