siRNA Details
Gene Symbol: SCP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007098.2 | NP_001007099.1 | 4 | 422 | ||
NM_001193599.1 | NP_001180528.1 | 4 | 482 | |||
NM_001193600.1 | NP_001180529.1 | 4 | 422 | |||
NM_001193617.1 | NP_001180546.1 | 4 | 496 | |||
NM_002979.4 | NP_002970.2 | 5 | 554 | |||
XM_005271103.4 | XP_005271160.1 | 5 | 497 | |||
XM_011541935.2 | XP_011540237.1 | 5 | 497 | |||
XM_017002045.1 | XP_016857534.1 | 5 | 497 | |||
GenBank mRNA | AB208789.1 | - | 5 | 506 | ||
AK294631.1 | - | 4 | 425 | |||
AK295214.1 | - | 4 | 439 | |||
AK308105.1 | - | 4 | 365 | |||
AK312856.1 | - | 5 | 505 | |||
BC067108.1 | - | 4 | 333 | 5176900 | ||
DC330171.1 | - | 5 | 538 | |||
M75883.1 | - | 5 | 407 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
