siRNA Details
Gene Symbol: BLNK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011539728.2 | XP_011538030.1 | 17 | 3575 | ||
XM_017016159.1 | XP_016871648.1 | 16 | 3507 |
Gene Symbol: CLEC2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004419.4 | NP_001004419.1 | 6 | 1591 | ||
NM_001197317.2 | NP_001184246.1 | 4 | 1398 | |||
NM_001197318.2 | NP_001184247.1 | 4 | 1405 | |||
NM_001197319.2 | NP_001184248.1 | 3 | 1294 | |||
NM_013269.5 | NP_037401.1 | 5 | 1509 | |||
NR_036693.2 | 4 | 1480 | ||||
GenBank mRNA | AK310554.1 | - | 4 | 1471 | ||
BC094859.1 | - | 1 | 386 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
