siRNA Details
Gene Symbol: ABCC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320032.1 | NP_001306961.1 | 20 | 3182 | ||
NM_005688.3 | NP_005679.2 | 20 | 3067 | |||
XM_005247058.4 | XP_005247115.1 | 20 | 4635 | |||
XM_005247059.4 | XP_005247116.1 | 20 | 3057 | |||
XM_011512314.1 | XP_011510616.1 | 20 | 2958 | |||
XM_011512315.1 | XP_011510617.1 | 20 | 3117 | |||
XM_017005492.1 | XP_016860981.1 | 17 | 4306 | |||
GenBank mRNA | AB005659.1 | - | 14 | 2144 | ||
AB019002.1 | - | 19 | 2933 | |||
AB209103.1 | - | 17 | 2575 | |||
AB209454.1 | - | 20 | 2994 | |||
AF104942.1 | - | 20 | 3006 | |||
AF146074.1 | - | 20 | 3077 | |||
AY196484.1 | - | 19 | 2893 | |||
BC140771.1 | - | 19 | 2943 | 9021639 | ||
BX647245.1 | - | 2 | 191 | |||
U83661.2 | - | 20 | 3000 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
