siRNA Details
Gene Symbol: SLC38A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001307936.1 | NP_001294865.1 | 7 | 911 | ||
NM_018976.4 | NP_061849.2 | 8 | 1027 | |||
GenBank mRNA | AB037803.2 | - | 7 | 587 | ||
AF259799.1 | - | 7 | 607 | |||
AF298897.1 | - | 8 | 938 | |||
AJ344099.1 | - | 8 | 939 | |||
AK001700.1 | - | 7 | 820 | |||
AK172784.1 | - | 1 | 34 | |||
AK291644.1 | - | 8 | 731 | |||
AK293858.1 | - | 1 | 142 | |||
BC040342.1 | - | 8 | 929 | 3874551 | ||
CR457267.1 | - | 4 | 287 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
