siRNA Details
Gene Symbol: PDE9A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001573.1 | NP_001001573.1 | 2 | 151 | ||
NM_001001581.1 | NP_001001581.1 | 3 | 195 | |||
NM_001001582.1 | NP_001001582.1 | 4 | 273 | |||
NM_001001583.1 | NP_001001583.1 | 4 | 266 | |||
NM_001001585.1 | NP_001001585.1 | 4 | 273 | |||
NM_002606.2 | NP_002597.1 | 5 | 344 | |||
XM_011529598.2 | XP_011527900.1 | 5 | 266 | |||
XM_011529600.2 | XP_011527902.1 | 5 | 317 | |||
XM_017028367.1 | XP_016883856.1 | 5 | 317 | |||
GenBank mRNA | AF048837.1 | - | 5 | 341 | ||
AF067223.1 | - | 5 | 293 | |||
AK314679.1 | - | 5 | 317 | |||
AY196302.1 | - | 2 | 124 | |||
AY196310.1 | - | 3 | 168 | |||
AY196311.1 | - | 4 | 246 | |||
AY196312.1 | - | 4 | 239 | |||
AY196314.1 | - | 4 | 246 | |||
KU178247.1 | - | 5 | 281 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
