siRNA Details
Gene Symbol: MRC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002438.3 | NP_002429.1 | 11 | 1827 | ||
GenBank mRNA | AK308132.1 | - | 11 | 1829 | ||
AK308167.1 | - | 11 | 1823 | |||
BC146838.1 | - | 11 | 1750 | 9056765 | ||
DQ663787.1 | - | 11 | 1704 | |||
J05550.1 | - | 11 | 1807 | |||
X55635.1 | - | 11 | 1780 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
