siRNA Details
Gene Symbol: CKMT1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015001.2 | NP_001015001.1 | 3, 4 | 836 | ||
NM_001321926.1 | NP_001308855.1 | 2, 3 | 704 | |||
NM_001321927.1 | NP_001308856.1 | 3, 4 | 929 | |||
NM_001321928.1 | NP_001308857.1 | 3, 4 | 690 | |||
NM_001321929.1 | NP_001308858.1 | 2, 3 | 732 | |||
NR_135856.1 | 2, 3 | 981 | ||||
XM_005254498.3 | XP_005254555.1 | 2, 3 | 455 | |||
XM_017022369.1 | XP_016877858.1 | 3, 4 | 761 | |||
XM_017022370.1 | XP_016877859.1 | 3, 4 | 686 | |||
XM_017022371.1 | XP_016877860.1 | 3, 4 | 1071 | |||
GenBank mRNA | AK223365.1 | - | 2, 3 | 516 | ||
AK295776.1 | - | 2, 3 | 580 | |||
AK316052.1 | - | 3, 4 | 380 | |||
AK316124.1 | - | 3, 4 | 787 | |||
AK316319.1 | - | 3, 4 | 690 | |||
BC001926.1 | - | 2, 3 | 521 | 2820642 | ||
BC006467.1 | - | 2, 3 | 447 | 3535486 | ||
BM923431.1 | - | 3, 4 | 755 | |||
BT006628.1 | - | 2, 3 | 344 |
Gene Symbol: CKMT1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020990.4 | NP_066270.1 | 3, 4 | 933 | ||
NR_135748.1 | 2, 3 | 982 | ||||
NR_135749.1 | 2, 3 | 1006 | ||||
NR_135750.1 | 3, 4 | 933 | ||||
NR_135751.1 | 2, 3 | 822 | ||||
XM_005254150.3 | XP_005254207.1 | 2, 3 | 455 | |||
XM_011521194.1 | XP_011519496.1 | 3, 4 | 1021 | |||
XM_011521195.2 | XP_011519497.1 | 3, 4 | 760 | |||
XM_011521196.1 | XP_011519498.1 | 3, 4 | 686 | |||
XM_011521197.2 | XP_011519499.1 | 2, 3 | 520 | |||
XM_011521198.1 | XP_011519500.1 | 3, 4 | 1205 | |||
XM_011521199.2 | XP_011519501.2 | 3, 4 | 1021 | |||
XM_017021902.1 | XP_016877391.1 | 3, 4 | 1205 | |||
GenBank mRNA | AK094322.1 | - | 3, 4 | 511 | ||
AK293685.1 | - | 2, 3 | 249 | |||
AK293790.1 | - | 2, 3 | 789 | |||
AK293939.1 | - | 2, 3 | 586 | |||
AK294002.1 | - | 2, 3 | 631 | |||
AK294063.1 | - | 2, 3 | 404 | |||
AK294735.1 | - | 3, 4 | 404 | |||
AK294908.1 | - | 2, 3 | 821 | |||
AK296271.1 | - | 1, 2 | 66 | |||
AK296472.1 | - | 2, 3 | 468 | |||
BC108652.1 | - | 2, 3 | 456 | 6190035 | ||
BC121001.1 | - | 2, 3 | 520 | 40116549 | ||
BC121002.1 | - | 2, 3 | 520 | 40116552 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
