siRNA Details
Gene Symbol: PCBD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000281.3 | NP_000272.1 | 4 | 745 | ||
NM_001289797.1 | NP_001276726.1 | 4 | 891 | |||
GenBank mRNA | AF082858.1 | - | 4 | 566 | ||
BC006324.2 | - | 4 | 510 | 4138350 | ||
L41559.1 | - | 4 | 513 | |||
M83742.1 | - | 3 | 494 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
