siRNA Details
Gene Symbol: MYL6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199629.1 | NP_001186558.1 | 8 | 1025 | ||
NM_002475.4 | NP_002466.1 | 8 | 961 | |||
GenBank mRNA | BC012425.1 | - | 7 | 749 | 3862144 | |
BC014400.1 | - | 8 | 1025 | 3503030 | ||
M31211.1 | - | 7 | 729 | |||
X16434.1 | - | 7 | 714 |
