siRNA Details
Gene Symbol: METTL7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014033.3 | NP_054752.3 | 1 | 461 | ||
XM_006719332.1 | XP_006719395.1 | 2 | 815 | |||
GenBank mRNA | AF113007.1 | - | 1 | 135 | ||
AJ536233.1 | - | 1 | 183 | |||
AK023693.1 | - | 1 | 193 | |||
AK024409.1 | - | 1 | 226 | |||
AK225595.1 | - | 1 | 198 | |||
AK225689.1 | - | 1 | 197 | |||
AK293356.1 | - | 1 | 194 | |||
AK307071.1 | - | 1 | 201 | |||
AL050159.1 | - | 1 | 171 | |||
AY358797.1 | - | 1 | 229 | |||
BC004492.2 | - | 1 | 178 | 3689692 | ||
BC008180.2 | - | 1 | 179 | 2900478 | ||
BM805242.1 | - | 1 | 483 |
