siRNA Details
Gene Symbol: PGS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024419.4 | NP_077733.3 | 5 | 713 | ||
NR_110601.1 | 4 | 635 | ||||
NR_110602.1 | 4 | 597 | ||||
NR_111989.1 | 4 | 523 | ||||
XM_011525487.2 | XP_011523789.1 | 5 | 1088 | |||
XM_011525488.2 | XP_011523790.1 | 5 | 1088 | |||
XM_011525489.2 | XP_011523791.1 | 5 | 1088 | |||
XM_017025357.1 | XP_016880846.1 | 5 | 1088 | |||
XM_017025358.1 | XP_016880847.1 | 5 | 1088 | |||
XM_017025359.1 | XP_016880848.1 | 5 | 1088 | |||
XM_017025360.1 | XP_016880849.1 | 5 | 1088 | |||
XM_017025361.1 | XP_016880850.1 | 5 | 707 | |||
XM_017025362.1 | XP_016880851.1 | 5 | 707 | |||
XM_017025363.1 | XP_016880852.1 | 5 | 659 | |||
XM_017025364.1 | XP_016880853.1 | 4 | 609 | |||
XM_017025365.1 | XP_016880854.1 | 4 | 561 | |||
XM_017025366.1 | XP_016880855.1 | 5 | 707 | |||
GenBank mRNA | AK024529.1 | - | 4 | 491 | ||
AK225030.1 | - | 4 | 492 | |||
AK296882.1 | - | 5 | 687 | |||
AK297156.1 | - | 4 | 553 | |||
AK307874.1 | - | 5 | 689 | |||
AK309890.1 | - | 1 | 214 | |||
AK316147.1 | - | 5 | 691 | |||
AK316405.1 | - | 4 | 600 | |||
BC008903.2 | - | 5 | 661 | |||
BC015570.2 | - | 5 | 661 | |||
BC025951.1 | - | 5 | 646 | |||
BC035662.1 | - | 4 | 596 | |||
BC108732.1 | - | 5 | 677 | 6504169 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
